![(S)-(+)-Camptothecin ≥90% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/123/089/e944834d-8145-4adb-9040-27d6ac64aab8/640/e944834d-8145-4adb-9040-27d6ac64aab8.png)
(S)-(+)-Camptothecin
![(S)-(+)-Camptothecin ≥90% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/123/089/e944834d-8145-4adb-9040-27d6ac64aab8/640/e944834d-8145-4adb-9040-27d6ac64aab8.png)
Synonym(s):
Camptothecin, Camptotheca acuminata
Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-2 of 2