Skip to Content
MilliporeSigma
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid 98%

(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid

(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid 98%
Synonym(s):
Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing