Skip to Content
MilliporeSigma
Scopoletin phyproof® Reference Substance

Scopoletin

Scopoletin phyproof® Reference Substance
Synonym(s):
6-Methoxyumbelliferone, 6-Methylesculetin, 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin-6-methyl ether, Gelseminic acid, Murrayetin
Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4