Skip to Content
MilliporeSigma
Scopolamine Hydrobromide Pharmaceutical Secondary Standard; Certified Reference Material

Scopolamine hydrobromide

Scopolamine Hydrobromide Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(−)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide, Scopine tropate, Scopolamine hydrobromide
Empirical Formula (Hill Notation):
C17H21NO4 · HBr · 3H2O
CAS Number:
Molecular Weight:
438.31

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique