Skip to Content
MilliporeSigma
(S)-cis-Verbenol 95%

(S)-cis-Verbenol

(S)-cis-Verbenol 95%
Synonym(s):
(1S,2S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-ol
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing