Skip to Content
MilliporeSigma
SC-58125 ≥98% (HPLC)

SC-58125

SC-58125 ≥98% (HPLC)
Synonym(s):
5-(4-Fluorophenyl)-1-[4-(methylsulfonyl)phenyl]-3-(trifluoromethyl)pyrazole
Empirical Formula (Hill Notation):
C17H12N2SO2F4
CAS Number:
Molecular Weight:
384.35

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing