
S-Boc-2-mercapto-4,6-dimethylpyrimidine

Synonym(s):
O-tert-Butyl-S-(4,6-dimethyl-2-pyrimidinyl) thiocarbonate, Boc-S
Empirical Formula (Hill Notation):
C11H16N2O2S
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1