Skip to Content
MilliporeSigma
(S)-β-Hydroxy-γ-butyrolactone 96%

(S)-β-Hydroxy-γ-butyrolactone

(S)-β-Hydroxy-γ-butyrolactone 96%
Synonym(s):
(S)-4,5-Dihydro-4-hydroxy-2(3H)-furanone
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing