![Saxagliptin hydrate ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/409/920/2e5c4898-93bd-4e36-b78f-3638443877d0/640/2e5c4898-93bd-4e36-b78f-3638443877d0.png)
Saxagliptin hydrate
![Saxagliptin hydrate ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/409/920/2e5c4898-93bd-4e36-b78f-3638443877d0/640/2e5c4898-93bd-4e36-b78f-3638443877d0.png)
Synonym(s):
(S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11
Empirical Formula (Hill Notation):
C18H25N3O2 · H2O
CAS Number:
Molecular Weight:
333.43
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing