Skip to Content
MilliporeSigma
(S)-AMPA ≥98%

(S)-AMPA

(S)-AMPA ≥98%
Synonym(s):
(S)-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid
Empirical Formula (Hill Notation):
C7H10N2O4
CAS Number:
Molecular Weight:
186.17

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing