Skip to Content
MilliporeSigma
(S)-α-Methyl-4-nitrobenzylamine hydrochloride 97%

(S)-α-Methyl-4-nitrobenzylamine hydrochloride

(S)-α-Methyl-4-nitrobenzylamine hydrochloride 97%
Synonym(s):
(S)-1-(4-Nitrophenyl)ethylamine hydrochloride
Linear Formula:
O2NC6H4CH(CH3)NH2 · HCl
CAS Number:
Molecular Weight:
202.64

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing