![Salsalate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/373/032/0f644466-d46c-4ec7-8f2e-5675ca9658ed/640/0f644466-d46c-4ec7-8f2e-5675ca9658ed.png)
Salsalate
![Salsalate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/373/032/0f644466-d46c-4ec7-8f2e-5675ca9658ed/640/0f644466-d46c-4ec7-8f2e-5675ca9658ed.png)
Synonym(s):
2-(2-Hydroxybenzoyl)oxybenzoic acid, 2-Hydroxybenzoic acid 2-carboxyphenyl ester, disalicylic acid, salicyloxysalicylic acid, salicylsalicylic acid
Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing