Skip to Content
MilliporeSigma
Salmeterol xinafoate ≥98% (HPLC), solid

Salmeterol Xinafoate

Salmeterol xinafoate ≥98% (HPLC), solid
Synonym(s):
Salmeterol xinafoate, (±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate
Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique