Skip to Content
MilliporeSigma
(S)-(−)-α-Amino-γ-butyrolactone hydrobromide 99%

(S)-(−)-α-Amino-γ-butyrolactone hydrobromide

(S)-(−)-α-Amino-γ-butyrolactone hydrobromide 99%
Synonym(s):
L-Homoserine lactone hydrobromide
Empirical Formula (Hill Notation):
C4H7NO2 · HBr
CAS Number:
Molecular Weight:
182.02

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing