Skip to Content
MilliporeSigma
S-(5′-Adenosyl)-L-methionine chloride dihydrochloride ≥75%

S-(5′-Adenosyl)-L-methionine chloride dihydrochloride

S-(5′-Adenosyl)-L-methionine chloride dihydrochloride ≥75%
Synonym(s):
Active methionine, AdoMet, SAM chloride dihydrochloride
Empirical Formula (Hill Notation):
C15H23ClN6O5S · 2 HCl
CAS Number:
Molecular Weight:
507.82

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing