Skip to Content
MilliporeSigma
(S)-(−)-4-Isopropyl-2-oxazolidinone 99%

(S)-(−)-4-Isopropyl-2-oxazolidinone

(S)-(−)-4-Isopropyl-2-oxazolidinone 99%
Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing