Skip to Content
MilliporeSigma
(S)-(−)-4-Amino-2-hydroxybutyric acid 96%

(S)-(−)-4-Amino-2-hydroxybutyric acid

(S)-(−)-4-Amino-2-hydroxybutyric acid 96%
Linear Formula:
H2NCH2CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
119.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing