Skip to Content
MilliporeSigma
(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene 97%

(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene

(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene 97%
Synonym(s):
(S)-(−)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (S)-(−)-BINAP
Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing