Skip to Content
MilliporeSigma
(S)-2-(2,3-Bis(dicyclohexylamino)cyclopropenimine)-3-phenylpropan-1-ol hydrochloride ≥95%

(S)-2-(2,3-Bis(dicyclohexylamino)cyclopropenimine)-3-phenylpropan-1-ol hydrochloride

(S)-2-(2,3-Bis(dicyclohexylamino)cyclopropenimine)-3-phenylpropan-1-ol hydrochloride ≥95%
Empirical Formula (Hill Notation):
C36H55N3O · HCl
CAS Number:
Molecular Weight:
582.30

Products

Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1