Skip to Content
MilliporeSigma
(S)-(−)-1-(2-Naphthyl)ethylamine ≥99.0% (sum of enantiomers, GC)

(S)-(−)-1-(2-Naphthyl)ethylamine

(S)-(−)-1-(2-Naphthyl)ethylamine ≥99.0% (sum of enantiomers, GC)
Synonym(s):
(S)-(−)-α-Methyl-2-naphthalenemethylamine
Empirical Formula (Hill Notation):
C12H13N
CAS Number:
Molecular Weight:
171.24

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing