Skip to Content
MilliporeSigma
(S)-(−)-1,1′-Bi(2-naphthol) 99%

(S)-(−)-1,1′-Bi(2-naphthol)

(S)-(−)-1,1′-Bi(2-naphthol) 99%
Synonym(s):
(−)-2,2′-Dihydroxy-1,1′-dinaphthyl, (S)-(−)-1,1′-Binaphthalene-2,2′-diol, (S)-BINOL
Linear Formula:
HOC10H6C10H6OH
CAS Number:
Molecular Weight:
286.32

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing