Skip to Content
MilliporeSigma
Rotigotine hydrochloride European Pharmacopoeia (EP) Reference Standard

Rotigotine hydrochloride

Rotigotine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(−)-(S)-5,6,7,8-Tetrahydro-6-(propyl(2-(2-thienyl)ethyl)amino)-1-naphthol hydrochloride, (−)-N 0437, N 0923, Neupro, SPM 962
Empirical Formula (Hill Notation):
C19H25NOS·HCl
CAS Number:
Molecular Weight:
351.93

Products

Product Category
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
API Family
Special Grade
Showing 1-2 of 2