Skip to Content
MilliporeSigma
Rosuvastatin Impurity D certified reference material, pharmaceutical secondary standard

Rosuvastatin Impurity D

Rosuvastatin Impurity D certified reference material, pharmaceutical secondary standard
Synonym(s):
N-[4-(4-fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethenyl]-6-(1-methylethyl)pyrimidin-2-yl]-N-methylmethanesulfonamide
Empirical Formula (Hill Notation):
C22H26FN3O5S
CAS Number:
Molecular Weight:
463.52

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing