Skip to Content
MilliporeSigma
Risedronate Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material

Risedronate Related Compound C

Risedronate Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-(3-Pyridinyl)ethylidene]bis(phosphonic acid), 2-(Pyridin-3-yl)ethane-1,1-diyl]bis(phosphonic acid)
Empirical Formula (Hill Notation):
C7H11NO6P2
CAS Number:
Molecular Weight:
267.11

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade