Skip to Content
MilliporeSigma
Rhodblock 6 ≥98% (HPLC)

Rhodblock 6

Rhodblock 6 ≥98% (HPLC)
Synonym(s):
N-1H-Indazol-5-yl-cyclobutanecarboxamide
Empirical Formula (Hill Notation):
C12H13N3O
CAS Number:
Molecular Weight:
215.25

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing