Skip to Content
MilliporeSigma
R-(−)-Fluoxetine hydrochloride >98% (HPLC), solid

R-(−)-Fluoxetine hydrochloride

R-(−)-Fluoxetine hydrochloride >98% (HPLC), solid
Synonym(s):
(R)-N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine hydrochloride
Empirical Formula (Hill Notation):
C17H18NOF3 · HCl
CAS Number:
Molecular Weight:
345.79

Products

Product Category
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing