Skip to Content
MilliporeSigma
Resazurin sodium salt certified by the BSC

Resazurin sodium salt

Resazurin sodium salt certified by the BSC
Synonym(s):
7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
Empirical Formula (Hill Notation):
C12H6NNaO4
CAS Number:
Molecular Weight:
251.17
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing