Replacement 0.5 μm stainless steel frits for 2 mm I.D. TSKgel® HPLC Columns
Synonym(s):
TSKgel® Hardware and Accessories
Products
Product Category
Manufacturer
Product Line
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing