Skip to Content
MilliporeSigma
(R)-(−)-Epichlorohydrin 99%

(R)-(−)-Epichlorohydrin

(R)-(−)-Epichlorohydrin 99%
Synonym(s):
(R)-(−)-2-(Chloromethyl)oxirane
Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing