Skip to Content
MilliporeSigma
Reichstein’s substance S ≥98%

Reichstein’s substance S

Reichstein’s substance S ≥98%
Synonym(s):
11-Deoxy-17-hydroxycorticosterone, 11-Deoxycortisol, 17α,21-Dihydroxy-4-pregnene-3,20-dione, 17α-Hydroxycortexone, 17,21-Dihydroxy-4-pregnene-3,20-dione, 17,21-Dihydroxyprogesterone, 4-Pregnene-17α,21-diol-3,20-dione, Cortexolone, Reichstein’s Compound S
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing