Skip to Content
MilliporeSigma
Rebaudioside D phyproof® Reference Substance

Rebaudioside D

Rebaudioside D phyproof® Reference Substance
Synonym(s):
(4α)-13-[(O-β-D-Glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid 2-O-β-D-glucopyranosyl-β-D-glucopyranosyl ester
Empirical Formula (Hill Notation):
C50H80O28
CAS Number:
Molecular Weight:
1129.15

Products

Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing