Skip to Content
MilliporeSigma
R(+)-SCH-23390 hydrochloride ≥98% (HPLC), solid

R(+)-SCH-23390 hydrochloride

R(+)-SCH-23390 hydrochloride ≥98% (HPLC), solid
Synonym(s):
R(+)-7-Chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride
Empirical Formula (Hill Notation):
C17H18ClNO · HCl
CAS Number:
Molecular Weight:
324.24

Products

Product Category
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing