Skip to Content
MilliporeSigma
(R)-(+)-Propylene oxide 99%

(R)-(+)-Propylene oxide

(R)-(+)-Propylene oxide 99%
Synonym(s):
(R)-(+)-1,2-Epoxypropane, (R)-(+)-Methyloxirane
Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing