Skip to Content
MilliporeSigma
R-(−)-Deprenyl hydrochloride powder, ≥98% (HPLC)

R-(−)-Deprenyl hydrochloride

R-(−)-Deprenyl hydrochloride powder, ≥98% (HPLC)
Synonym(s):
(R)-(−)-N,α-Dimethyl-N-(2-propynyl)phenethylamine hydrochloride, R(−)-N-α-Dimethyl-N-2-propynyl-benzeneethanamine hydrochloride, Selegiline hydrochloride
Empirical Formula (Hill Notation):
C13H17N · HCl
CAS Number:
Molecular Weight:
223.74

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing