Skip to Content
MilliporeSigma
(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid 99%

(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid

(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid 99%
Synonym(s):
(+)-MTPA, Mosher’s acid
Linear Formula:
C6H5C(OCH3)(CF3)CO2H
CAS Number:
Molecular Weight:
234.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing