Skip to Content
MilliporeSigma
(R)-(+)-2-Chloropropionic acid 99%

(R)-(+)-2-Chloropropionic acid

(R)-(+)-2-Chloropropionic acid 99%
Linear Formula:
CH3CH(Cl)CO2H
CAS Number:
Molecular Weight:
108.52

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing