Skip to Content
MilliporeSigma
vibrant-m

Ranolazine Related Compound B

vibrant-m
Synonym(s):
N-(2,6-Dimethylphenyl)-2-[4-(2-hydroxy-3-phenoxypropyl)piperazin-1-yl]acetamide dihydrochloride monohydrate
Empirical Formula (Hill Notation):
C23H31N3O3 · 2HCl · H2O
Molecular Weight:
488.45

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade