Skip to Content
MilliporeSigma
Ranitidine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Ranitidine Related Compound A

Ranitidine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-{{{5-[(Dimethylamino)methyl]furan-2-yl}methyl}sulfanyl}ethanamine hemifumarate salt, 2-{{{5-[(Dimethylamino)methyl]-2-furanyl}methyl}thio}ethylamine hemifumarate salt, 5-{[(2-Aminoethyl)thio]methyl}-N,N-dimethyl- 2-furanmethanamine hemifumarate salt, Ranitidine diamine hemifumarate salt
Empirical Formula (Hill Notation):
C10H18N2OS · 0.5 C4H4O4
CAS Number:
Molecular Weight:
272.36

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique