Skip to Content
MilliporeSigma
Ranitidine hydrochloride British Pharmacopoeia (BP) Reference Standard

Ranitidine hydrochloride

Ranitidine hydrochloride British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C13H22N4O3S · HCl
CAS Number:
Molecular Weight:
350.86
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique