Skip to Content
MilliporeSigma
(R)-(−)-3-Chloro-1,2-propanediol 97%, optical purity ee: 98% (GLC)

(R)-(−)-3-Chloro-1,2-propanediol

(R)-(−)-3-Chloro-1,2-propanediol 97%, optical purity ee: 98% (GLC)
Synonym(s):
(R)-α-Glycerol chlorohydrin
Linear Formula:
ClCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
110.54

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing