Skip to Content
MilliporeSigma
(R)-3-(Boc-amino)piperidine 97%

(R)-3-(Boc-amino)piperidine

(R)-3-(Boc-amino)piperidine 97%
Synonym(s):
tert-Butyl (R)-piperidin-3-ylcarbamate, (R)-3-tert-Butoxycarbonylaminopiperidine
Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing