Skip to Content
MilliporeSigma
Quinoline analytical standard

Quinoline

Quinoline analytical standard
Synonym(s):
1-Benzazine, 2,3-Benzopyridine, Quinoline
Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing