Skip to Content
MilliporeSigma
Quinchlorac PESTANAL®, analytical standard

Quinchlorac

Quinchlorac PESTANAL®, analytical standard
Synonym(s):
3,7-Dichloro-8-quinolinecarboxylic acid
Empirical Formula (Hill Notation):
C10H5Cl2NO2
CAS Number:
Molecular Weight:
242.06
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing