Skip to Content
MilliporeSigma
Pyridinium tribromide technical grade, 90%

Pyridinium tribromide

Pyridinium tribromide technical grade, 90%
Synonym(s):
Pyridine hydrobromide perbromide, Pyridine perbromide hydrobromide, Pyridinium bromide perbromide
Empirical Formula (Hill Notation):
C5H5N·HBr·Br2
CAS Number:
Molecular Weight:
319.82
EC Number:

Products

Formula Weight
Physical Form
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing