Skip to Content
MilliporeSigma
Pyrene BCR®, certified reference material

Pyrene

Pyrene BCR®, certified reference material
Synonym(s):
Benzo[def]phenanthrene, Pyrene
Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique