Skip to Content
MilliporeSigma
PUGNAc ≥95% (HPLC)

PUGNAc

PUGNAc ≥95% (HPLC)
Synonym(s):
O-(2-Acetamido-2-deoxy-D-glucopyranosylidenamino) N-phenylcarbamate
Empirical Formula (Hill Notation):
C15H19N3O7
CAS Number:
Molecular Weight:
353.33

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing