Skip to Content
MilliporeSigma
p-Tolylboronic acid 97%

p-Tolylboronic acid

p-Tolylboronic acid 97%
Synonym(s):
(p-Methylphenyl)boronic acid, 4-Methylbenzeneboronic acid, 4-Methylphenylboronic acid, 4-Tolueneboronic acid, 4-Tolylboronic acid, p-Tolueneboronic acid, NSC 62870, p-Methylbenzeneboronic acid
Linear Formula:
CH3C6H4B(OH)2
CAS Number:
Molecular Weight:
135.96

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing