Skip to Content
MilliporeSigma
p-Toluenesulfonyl cyanide technical grade, 95%

p-Toluenesulfonyl cyanide

p-Toluenesulfonyl cyanide technical grade, 95%
Synonym(s):
p-Tolylsulfonyl cyanide, Tosyl cyanide
Linear Formula:
CH3C6H4SO2CN
CAS Number:
Molecular Weight:
181.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing