Skip to Content
MilliporeSigma
Pterostilbene ≥97% (HPLC), solid

Pterostilbene

Pterostilbene ≥97% (HPLC), solid
Synonym(s):
3,5-Dimethoxy-4′-hydroxystilbene, 4-[(1E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
Empirical Formula (Hill Notation):
C16H16O3
CAS Number:
Molecular Weight:
256.30

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing