Skip to Content
MilliporeSigma
Pteroic acid ≥93%

Pteroic acid

Pteroic acid ≥93%
Empirical Formula (Hill Notation):
C14H12N6O3
CAS Number:
Molecular Weight:
312.28

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing